Tisk…

3-Cyclohexyl-3-oxopropanenitrile 95%

Dodavatel: Apollo Scientific

OR930579-1G OR930579-250MG
APOSOR930579-1GEA 22950 CZK
APOSOR930579-1G APOSOR930579-250MG
3-Cyclohexyl-3-oxopropanenitrile 95%
3-Cyclohexyl-3-oxopropanenitrile
Vzorec: C₉H₁₃NO
MW: 151,206 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07700128
CAS číslo: 62455-70-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR