2-Amino-3-bromobenzaldehyde 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR930475-1G
OR930475-5G
APOSOR930475-1GEA
1410
CZK
APOSOR930475-1G
APOSOR930475-5G
2-Amino-3-bromobenzaldehyde 98%
2-Amino-3-bromobenzaldehyde
Vzorec:
C₇H₆BrNO MW: 200,033 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD18389339 CAS číslo: 145123-24-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...