Tisk…

2-Amino-cyclopent-1-enecarboxylic acid methyl ester 95%

Dodavatel: Apollo Scientific

OR930354-10G OR930354-1G OR930354-5G
APOSOR930354-10GEA 14680 CZK
APOSOR930354-10G APOSOR930354-1G APOSOR930354-5G
2-Amino-cyclopent-1-enecarboxylic acid methyl ester 95%
2-Amino-cyclopent-1-enecarboxylic acid methyl ester
Vzorec: C₇H₁₁NO₂
MW: 141.168 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19204033
CAS číslo: 52909-60-1

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR