1-Acetyl-1H-benzotriazole 95%
Dodavatel: Apollo Scientific
OR930339-1G
OR930339-5G
APOSOR930339-1GEA
1740
CZK
APOSOR930339-1G
APOSOR930339-5G
1-Acetyl-1H-benzotriazole 95%
1-Acetyl-1H-benzotriazole
Vzorec:
C₈H₇N₃O Storage Temperature: Ambient |
MDL Number:
MFCD00461288 CAS číslo: 18773-93-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...