1H-Benzo[d]azepin-2(3H)-one 95%
Dodavatel: Apollo Scientific
OR930300-1G
OR930300-250MG
APOSOR930300-1GEA
5960
CZK
APOSOR930300-1G
APOSOR930300-250MG
1H-Benzo[d]azepin-2(3H)-one 95%
1H-Benzo[d]azepin-2(3H)-one
Vzorec:
C₁₀H₉NO MW: 159,185 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01101239 CAS číslo: 19301-09-8 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...