Tisk…

2-(Sec-butylthio)pyrimidine-5-carbaldehyde 95%

Dodavatel: Apollo Scientific

OR930245-1G OR930245-5G
APOSOR930245-1GEA 4770 CZK
APOSOR930245-1G APOSOR930245-5G
2-(Sec-butylthio)pyrimidine-5-carbaldehyde 95%
2-(Sec-butylthio)pyrimidine-5-carbaldehyde
Vzorec: C₉H₁₂N₂OS
MW: 196,269 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08752993
CAS číslo: 915920-24-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR