2-(4-Hydroxyphenyl)-2-methylpropanenitrile 95%
Dodavatel: Apollo Scientific
OR930226-1G
OR930226-250MG
APOSOR930226-1GEA
1370
CZK
APOSOR930226-1G
APOSOR930226-250MG
2-(4-Hydroxyphenyl)-2-methylpropanenitrile 95%
2-(4-Hydroxyphenyl)-2-methylpropanenitrile
Vzorec:
C₁₀H₁₁NO MW: 161,2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11100986 CAS číslo: 55770-61-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...