2,3-Dihydro-4-benzofuranamine 95%
Dodavatel: Apollo Scientific
OR930204-1G
OR930204-250MG
APOSOR930204-1GEA
3870
CZK
APOSOR930204-1G
APOSOR930204-250MG
2,3-Dihydro-4-benzofuranamine 95%
2,3-Dihydro-4-benzofuranamine
Vzorec:
C₈H₉NO Storage Temperature: Ambient |
MDL Number:
MFCD06659470 CAS číslo: 61090-37-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...