1-(1-Propyl-1H-1,2,4-triazol-5-yl)ethanamine 95%
Dodavatel: Apollo Scientific
OR930182-1G
OR930182-250MG
APOSOR930182-1GEA
24600
CZK
APOSOR930182-1G
APOSOR930182-250MG
1-(1-Propyl-1H-1,2,4-triazol-5-yl)ethanamine 95%
1-(1-Propyl-1H-1,2,4-triazol-5-yl)ethanamine
Vzorec:
C₇H₁₄N₄ Storage Temperature: Ambient |
MDL Number:
MFCD11053902 CAS číslo: 1060817-05-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...