Tisk…

1-(3-Pyridin-3-ylisoxazol-5-yl)methanamine 95%

Dodavatel: Apollo Scientific

OR930141-1G OR930141-250MG
APOSOR930141-1GEA 25790 CZK
APOSOR930141-1G APOSOR930141-250MG
1-(3-Pyridin-3-ylisoxazol-5-yl)methanamine 95%
1-(3-Pyridin-3-ylisoxazol-5-yl)methanamine
Vzorec: C₉H₉N₃O
MW: 175,187 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08059814
CAS číslo: 543713-43-5
UN: 3259
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR