Tisk…

2-Chloro-5-methoxypyrazine 98%

Dodavatel: Apollo Scientific

OR930097-1G OR930097-250MG
APOSOR930097-1GEA 4630 CZK
APOSOR930097-1G APOSOR930097-250MG
2-Chloro-5-methoxypyrazine 98%
2-Chloro-5-methoxypyrazine
Vzorec: C₅H₅ClN₂O
MW: 144,559 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09607702
CAS číslo: 33332-31-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR