2-Bromo-5-methylphenol 98%
Dodavatel: Apollo Scientific
OR930096-25G
APOSOR930096-25GEA
3410
CZK
APOSOR930096-25G
2-Bromo-5-methylphenol 98%
2-Bromo-5-methylphenol
Vzorec:
C₇H₇BrO MW: 187,034 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11100989 CAS číslo: 14847-51-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...