Tisk…

Allantoic acid

Dodavatel: Apollo Scientific

OR930085-100MG OR930085-1G OR930085-250MG
APOSOR930085-100MGEA 8870 CZK
APOSOR930085-100MG APOSOR930085-1G APOSOR930085-250MG
Allantoic acid
Allantoic acid
Vzorec: C₄H₈N₄O₄
Storage Temperature: Ambient
MDL Number: MFCD00007949
CAS číslo: 99-16-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR