Tisk…

6-Chloro-4-methoxypyridazin-3-amine 96%

Dodavatel: Apollo Scientific

OR930059-1G OR930059-250MG
APOSOR930059-1GEA 6420 CZK
APOSOR930059-1G APOSOR930059-250MG
6-Chloro-4-methoxypyridazin-3-amine 96%
6-Chloro-4-methoxypyridazin-3-amine
Vzorec: C₅H₆ClN₃O
MW: 159,574 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20702054
CAS číslo: 808770-39-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR