Tisk…

Benzyl 3-hydroxypropionate 98%

Dodavatel: Apollo Scientific

Synonyms: Benzyl 3-Hydroxypropionate

OR929982-1G OR929982-5G
APOSOR929982-1GEA 2200 CZK
APOSOR929982-1G APOSOR929982-5G
Benzyl 3-hydroxypropionate 98%
Benzyl 3-hydroxypropionate
Vzorec: C₁₀H₁₂O₃
MW: 180,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00029657
CAS číslo: 14464-10-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR