Tisk…

3-Aminophenethyl alcohol 97%

Dodavatel: Apollo Scientific

Synonyms: 2-(3-Aminophenyl)ethanol

OR929971-1G OR929971-5G
APOSOR929971-1GEA 2940 CZK
APOSOR929971-1G APOSOR929971-5G
3-Aminophenethyl alcohol 97%
3-Aminophenethyl alcohol
Vzorec: C₈H₁₁NO
MW: 137,179 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02683213
CAS číslo: 52273-77-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR