Tisk…

Dimethyl(4-chlorophenyl)silanol 95%

Dodavatel: Apollo Scientific

OR929950-100MG OR929950-1G OR929950-250MG
APOSOR929950-100MGEA 4140 CZK
APOSOR929950-100MG APOSOR929950-1G APOSOR929950-250MG
Dimethyl(4-chlorophenyl)silanol 95%
Dimethyl(4-chlorophenyl)silanol
Vzorec: C₈H₁₁ClOSi
Storage Temperature: Ambient
MDL Number: MFCD09266144
CAS číslo: 18246-04-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR