Tisk…

4-Chloro-2-[(methylamino)methyl]phenol 95%

Dodavatel: Apollo Scientific

OR929938-1G OR929938-5G
APOSOR929938-1GEA 3580 CZK
APOSOR929938-1G APOSOR929938-5G
4-Chloro-2-[(methylamino)methyl]phenol 95%
4-Chloro-2-[(methylamino)methyl]phenol
Vzorec: C₈H₁₀ClNO
Storage Temperature: Ambient
MDL Number: MFCD09044328
CAS číslo: 38926-77-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR