Tisk…

1-(2-Chloroethyl)-5-methyl-1H-pyrazole 95%

Dodavatel: Apollo Scientific

OR929874-1G OR929874-250MG
APOSOR929874-1GEA 17360 CZK
APOSOR929874-1G APOSOR929874-250MG
1-(2-Chloroethyl)-5-methyl-1H-pyrazole 95%
1-(2-Chloroethyl)-5-methyl-1H-pyrazole
Vzorec: C₆H₉ClN₂
MW: 144,602 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02855989
CAS číslo: 96450-54-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR