Tisk…

1-Methyl-piperidine-4-carbonitrile 95%

Dodavatel: Apollo Scientific

Synonyms: 1-Methylpiperidine-4-carbonitrile

OR929855-1G
APOSOR929855-1GEA 2940 CZK
APOSOR929855-1G
1-Methyl-piperidine-4-carbonitrile 95%
1-Methyl-piperidine-4-carbonitrile
Vzorec: C₇H₁₂N₂
MW: 124,184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06658487
CAS číslo: 20691-92-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR