Tisk…

(4-Ethoxy-phenyl)-oxo-acetaldehyde 95%

Dodavatel: Apollo Scientific

OR929814-1G OR929814-5G
APOSOR929814-1GEA 3940 CZK
APOSOR929814-1G APOSOR929814-5G
(4-Ethoxy-phenyl)-oxo-acetaldehyde 95%
(4-Ethoxy-phenyl)-oxo-acetaldehyde
Vzorec: C₁₀H₁₀O₃
MW: 178,185 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08448031
CAS číslo: 14333-52-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR