Tisk…
5-Methylthiophen-3-amine hydrochloride 97%
Dodavatel: Apollo Scientific
OR929810-1G OR929810-250MG
APOSOR929810-1GEA 23680 CZK
APOSOR929810-1G APOSOR929810-250MG
5-Methylthiophen-3-amine hydrochloride 97%
5-Methylthiophen-3-amine hydrochloride
Vzorec: C₅H₇NS·ClH
MW: 149,642 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23703140
CAS číslo: 24030-01-1