Tisk…

2-[(Methylamino)methyl]benzyl alcohol 95%

Dodavatel: Apollo Scientific

Synonyms: 2-[(Methylamino)methyl]benzyl Alcohol

OR929723-1G OR929723-250MG
APOSOR929723-1GEA 12400 CZK
APOSOR929723-1G APOSOR929723-250MG
2-[(Methylamino)methyl]benzyl alcohol 95%
2-[(Methylamino)methyl]benzyl alcohol
Vzorec: C₉H₁₃NO
MW: 151.206 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09035678
CAS číslo: 57685-24-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR