Tisk…
3-(2-Pyridyl)-1-propanol hydrochloride 96%
Dodavatel: Apollo Scientific
OR929645-25G OR929645-5G
APOSOR929645-25GEA 9330 CZK
APOSOR929645-25G APOSOR929645-5G
3-(2-Pyridyl)-1-propanol hydrochloride 96%
3-(2-Pyridyl)-1-propanol hydrochloride
Vzorec: C₈H₁₁NO·ClH
MW: 173,64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23381121
CAS číslo: 52225-87-3