Tisk…

2-Methyl-3H-quinazolin-4-one hydrochloride 95%

Dodavatel: Apollo Scientific

OR929504-10G OR929504-1G
APOSOR929504-10GEA 17590 CZK
APOSOR929504-10G APOSOR929504-1G
2-Methyl-3H-quinazolin-4-one hydrochloride 95%
2-Methyl-3H-quinazolin-4-one hydrochloride
Vzorec: C₉H₈N₂O·ClH
MW: 196,634 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22570184
CAS číslo: 29378-39-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR