Cyclopropyl(pyridin-2-yl)methanone 95%
Dodavatel: Apollo Scientific
OR929500-1G
OR929500-250MG
APOSOR929500-1GEA
10350
CZK
APOSOR929500-1G
APOSOR929500-250MG
Cyclopropyl(pyridin-2-yl)methanone 95%
Cyclopropyl(pyridin-2-yl)methanone
Vzorec:
C₉H₉NO MW: 147,174 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07699200 CAS číslo: 57276-28-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...