Tisk…

Imidazo[1,2-a]pyridin-2-ol 95+%

Dodavatel: Apollo Scientific
Varování

OR929487-1G OR929487-250MG
APOSOR929487-1GEA 4270 CZK
APOSOR929487-1G APOSOR929487-250MG
Imidazo[1,2-a]pyridin-2-ol 95+%
Imidazo[1,2-a]pyridin-2-ol
Vzorec: C₇H₆N₂O
Storage Temperature: Ambient
MDL Number: MFCD09994219
CAS číslo: 112566-20-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR