3-(Hydroxymethyl)-4-propoxybenzaldehyde 95%
Dodavatel: Apollo Scientific
OR929438-1G
OR929438-250MG
APOSOR929438-1GEA
19440
CZK
APOSOR929438-1G
APOSOR929438-250MG
3-(Hydroxymethyl)-4-propoxybenzaldehyde 95%
3-(Hydroxymethyl)-4-propoxybenzaldehyde
Vzorec:
C₁₁H₁₄O₃ Storage Temperature: Ambient |
MDL Number:
MFCD08753026 CAS číslo: 915921-81-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...