Methyl 1H-pyrrolo[2,3-c]pyridine-3-carboxylate 95%
Dodavatel: Apollo Scientific
OR929431-1G
OR929431-250MG
APOSOR929431-1GEA
4100
CZK
APOSOR929431-1G
APOSOR929431-250MG
Methyl 1H-pyrrolo[2,3-c]pyridine-3-carboxylate 95%
Methyl 1H-pyrrolo[2,3-c]pyridine-3-carboxylate
Vzorec:
C₉H₈N₂O₂ MW: 176,172 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16036494 CAS číslo: 108128-12-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...