Tisk…

1-Ethyl-5-methyl-1H-pyrrole-2-carbaldehyde 95%

Dodavatel: Apollo Scientific

OR929417-1G OR929417-5G
APOSOR929417-1GEA 4770 CZK
APOSOR929417-1G APOSOR929417-5G
1-Ethyl-5-methyl-1H-pyrrole-2-carbaldehyde 95%
1-Ethyl-5-methyl-1H-pyrrole-2-carbaldehyde
Vzorec: C₈H₁₁NO
MW: 137,179 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12827513
CAS číslo: 39741-43-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR