Tisk…

N,N-Dimethylglycine methyl ester hydrochloride 95%

Dodavatel: Apollo Scientific

OR929353-1G OR929353-5G
APOSOR929353-1GEA 7250 CZK
APOSOR929353-1G APOSOR929353-5G
N,N-Dimethylglycine methyl ester hydrochloride 95%
N,N-Dimethylglycine methyl ester hydrochloride
Vzorec: C₅H₁₁NO₂·ClH
MW: 153,607 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08062582
CAS číslo: 1954-58-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR