Tisk…
Methyl 3-ethyl-2-hydroxybenzoate 95%
Dodavatel: Apollo Scientific
OR929281-1G OR929281-250MG
APOSOR929281-1GEA 17990 CZK
APOSOR929281-1G APOSOR929281-250MG
Methyl 3-ethyl-2-hydroxybenzoate 95%
Methyl 3-ethyl-2-hydroxybenzoate
Vzorec: C₁₀H₁₂O₃
MW: 180.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23381122
CAS číslo: 75871-40-8