Tisk…

4,5-Dimethyl-1,3-thiazole-2-carbaldehyde 98%

Dodavatel: Apollo Scientific

Synonyms: 4,5-Dimethylthiazole-2-carbaldehyde

OR929263-1G OR929263-5G
APOSOR929263-1GEA 1330 CZK
APOSOR929263-1G APOSOR929263-5G
4,5-Dimethyl-1,3-thiazole-2-carbaldehyde 98%
4,5-Dimethyl-1,3-thiazole-2-carbaldehyde
Vzorec: C₆H₇NOS
MW: 141,191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11044203
CAS číslo: 74531-15-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR