Tisk…

(E)-3-(2-Pyridinyl)-2-propen-1-ol 95%

Dodavatel: Apollo Scientific

OR929262-1G OR929262-250MG
APOSOR929262-1GEA 17990 CZK
APOSOR929262-1G APOSOR929262-250MG
(E)-3-(2-Pyridinyl)-2-propen-1-ol 95%
(E)-3-(2-Pyridinyl)-2-propen-1-ol
Vzorec: C₈H₉NO
MW: 135,163 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18384384
CAS číslo: 131610-03-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR