Tisk…

(S)-tert-Butanethiosulphinate 95%

Dodavatel: Apollo Scientific

Synonyms: (S)-S-tert-Butyl 2-methylpropane-2-sulphinothioate

OR929219-1G OR929219-250MG OR929219-5G
APOSOR929219-1GEA 3810 CZK
APOSOR929219-1G APOSOR929219-250MG APOSOR929219-5G
(S)-tert-Butanethiosulphinate 95%
(S)-tert-Butanethiosulphinate
Vzorec: C₈H₁₈OS₂
MW: 194,358 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09028107
CAS číslo: 60011-16-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR