Tisk…

2,3-Dihydrocinnolin-4(1H)-one hydrochloride 95%

Dodavatel: Apollo Scientific

OR929099-1G OR929099-250MG
APOSOR929099-1GEA 17990 CZK
APOSOR929099-1G APOSOR929099-250MG
2,3-Dihydrocinnolin-4(1H)-one hydrochloride 95%
2,3-Dihydrocinnolin-4(1H)-one hydrochloride
Vzorec: C₈H₈N₂O·ClH
MW: 184,623 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00156669
CAS číslo: 137195-33-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR