Tisk…

tert-Butyl 2-(4-bromophenyl)acetate 97%

Dodavatel: Apollo Scientific

Synonyms: tert-Butyl 2-(4-Bromophenyl)acetate

OR929080-1G OR929080-250MG
APOSOR929080-1GEA 2240 CZK
APOSOR929080-1G APOSOR929080-250MG
tert-Butyl 2-(4-bromophenyl)acetate 97%
tert-Butyl 2-(4-bromophenyl)acetate
Vzorec: C₁₂H₁₅BrO₂
MW: 271,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11975535
CAS číslo: 33155-58-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR