Tisk…

(1-Ethylpropyl)methylamine 95%

Dodavatel: Apollo Scientific

OR928914-1G OR928914-5G
APOSOR928914-1GEA 6220 CZK
APOSOR928914-1G APOSOR928914-5G
(1-Ethylpropyl)methylamine 95%
(1-Ethylpropyl)methylamine
Vzorec: C₆H₁₅N
Storage Temperature: Ambient
MDL Number: MFCD09046900
CAS číslo: 52317-98-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR