2-Chloro-1-ethyl-1H-imidazole 95%
Dodavatel: Apollo Scientific
OR928703-1G
OR928703-500MG
APOSOR928703-1GEA
16340
CZK
APOSOR928703-1G
APOSOR928703-500MG
2-Chloro-1-ethyl-1H-imidazole 95%
2-Chloro-1-ethyl-1H-imidazole
Vzorec:
C₅H₇ClN₂ MW: 130,575 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10699124 CAS číslo: 946061-13-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...