4-Chloro-6-hydroxynicotinic acid 95%
Dodavatel: Apollo Scientific
OR928671-1G
OR928671-250MG
APOSOR928671-1GEA
8830
CZK
APOSOR928671-1G
APOSOR928671-250MG
4-Chloro-6-hydroxynicotinic acid 95%
4-Chloro-6-hydroxynicotinic acid
Vzorec:
C₆H₄ClNO₃ MW: 173,554 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13189223 CAS číslo: 73038-85-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...