Tisk…

4-Methyl-2,3-dihydro-1H-indole 95%

Dodavatel: Apollo Scientific

Synonyms: 4-Methylindoline

OR928566-1G OR928566-250MG OR928566-5G
APOSOR928566-1GEA 6850 CZK
APOSOR928566-1G APOSOR928566-250MG APOSOR928566-5G
4-Methyl-2,3-dihydro-1H-indole 95%
4-Methyl-2,3-dihydro-1H-indole
Vzorec: C₉H₁₁N
MW: 133,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08437632
CAS číslo: 62108-16-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR