Tisk…

4-Methyl-cinnamaldehyde 95%

Dodavatel: Apollo Scientific

Synonyms: (E)-3-(p-Tolyl)acrylaldehyde

OR928560-10G
APOSOR928560-10GEA 662 CZK
APOSOR928560-10G
4-Methyl-cinnamaldehyde 95%
4-Methyl-cinnamaldehyde
Vzorec: C₁₀H₁₀O
MW: 146,186 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08460295
CAS číslo: 56578-35-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR