Tisk…

3-(Chloromethyl)pyridazine hydrochloride

Dodavatel: Apollo Scientific

OR928537-1G
APOSOR928537-1GEA 10550 CZK
APOSOR928537-1G
3-(Chloromethyl)pyridazine hydrochloride
3-(Chloromethyl)pyridazine hydrochloride
Vzorec: C₅H₅ClN₂·ClH
MW: 165,021 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19705404
CAS číslo: 27349-66-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR