Tisk…

5-Chloro-2,3-dimethylpyrazine 98%

Dodavatel: Apollo Scientific

OR928504-1G OR928504-250MG
APOSOR928504-1GEA 3310 CZK
APOSOR928504-1G APOSOR928504-250MG
5-Chloro-2,3-dimethylpyrazine 98%
5-Chloro-2,3-dimethylpyrazine
Vzorec: C₆H₇ClN₂
MW: 142,586 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13189517
CAS číslo: 59489-32-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR