Tisk…

6-(Piperidin-1-yl)pyridine-2-carboxylic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Carboxy-6-(piperidin-1-yl)pyridine, 1-(6-Carboxypyridin-2-yl)piperidine, 6-(Piperidin-1-yl)picolinic acid

OR9285-1G OR9285-250MG
APOSOR9285-1GEA 9200 CZK
APOSOR9285-1G APOSOR9285-250MG
6-(Piperidin-1-yl)pyridine-2-carboxylic acid 97%
6-(Piperidin-1-yl)pyridine-2-carboxylic acid
Vzorec: C₁₁H₁₄N₂O₂
MW: 206,241 g/mol
Bod tání: 97,5…101.5 °C
Storage Temperature: Ambient
MDL Number: MFCD08435860
CAS číslo: 868755-50-4
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR