Tisk…

3,4-Dimethoxy-2-methylbenzoic acid 95%

Dodavatel: Apollo Scientific

OR928435-1G OR928435-250MG
APOSOR928435-1GEA 9330 CZK
APOSOR928435-1G APOSOR928435-250MG
3,4-Dimethoxy-2-methylbenzoic acid 95%
3,4-Dimethoxy-2-methylbenzoic acid
Vzorec: C₁₀H₁₂O₄
MW: 196.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09701444
CAS číslo: 5722-94-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR