Tisk…

Methyl 2-(1H-pyrrolo[2,3-b]pyridin-1-yl)acetate 95%

Dodavatel: Apollo Scientific

OR928397-1G OR928397-250MG
APOSOR928397-1GEA 18420 CZK
APOSOR928397-1G APOSOR928397-250MG
Methyl 2-(1H-pyrrolo[2,3-b]pyridin-1-yl)acetate 95%
Methyl 2-(1H-pyrrolo[2,3-b]pyridin-1-yl)acetate
Vzorec: C₁₀H₁₀N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09834894
CAS číslo: 172647-94-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR