Tisk…

7-Bromo-2-methyloxazolo[4,5-c]pyridine 97%

Dodavatel: Apollo Scientific
Varování

OR928352-100MG OR928352-250MG
APOSOR928352-100MGEA 3310 CZK
APOSOR928352-100MG APOSOR928352-250MG
7-Bromo-2-methyloxazolo[4,5-c]pyridine 97%
7-Bromo-2-methyloxazolo[4,5-c]pyridine
Vzorec: C₇H₅BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD08692342
CAS číslo: 116081-17-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR