Tisk…

4-Ethyl-1H-pyrrole-3-carboxylic acid methyl ester 95%

Dodavatel: Apollo Scientific

OR928304-1G OR928304-250MG
APOSOR928304-1GEA 22950 CZK
APOSOR928304-1G APOSOR928304-250MG
4-Ethyl-1H-pyrrole-3-carboxylic acid methyl ester 95%
4-Ethyl-1H-pyrrole-3-carboxylic acid methyl ester
Vzorec: C₈H₁₁NO₂
MW: 153,178 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11875848
CAS číslo: 1260827-11-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR