3-Cyano-6-hydroxy-2-methoxypyridine 95%
Dodavatel: Apollo Scientific
OR928260-1G
OR928260-5G
APOSOR928260-1GEA
6220
CZK
APOSOR928260-1G
APOSOR928260-5G
3-Cyano-6-hydroxy-2-methoxypyridine 95%
3-Cyano-6-hydroxy-2-methoxypyridine
Vzorec:
C₇H₆N₂O₂ MW: 150,135 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11044304 CAS číslo: 130747-60-3 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...